Linolsyre for syntese, Sigma-Aldrich®

Leverandør: Merck


M-Clarity™ quality level = MQ200

8.43483.0025 8.43483.0100 8.43483.8100
8.43483.0025EA 3140 NOK
8.43483.0025 8.43483.0100 8.43483.8100
Linolsyre for syntese, Sigma-Aldrich®
Linolsyre
Formel: C₁₈H₃₂O₂
Kokepunkt: 360 °C (1013 hPa)
Smeltepunkt: –5 °C
Tetthet: 0,9 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064241
CAS-nummer: 60-33-3
EINECS: 200-470-9
Merck Indeks: 13,05527

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) practically insoluble
Melting Point -5 °C
Molar Mass 280.45 g/mol
Boiling Point 230 °C (21 hPa)
Flash Point 110 °C
Density 0.903 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR