2'-Methylacetoacetanilide ≥98% (by HPLC) for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 2-Methylacetoacetanilide, 2-Acetoacetotoluidide

M-Clarity™ quality level = MQ200

8.43847.0100
8.43847.0100EA 380 NOK
8.43847.0100
2'-Methylacetoacetanilide ≥98% (by HPLC) for synthesis, Sigma-Aldrich®
2'-Methylacetoacetanilide
Formel: C₁₁H₁₃NO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008782
CAS-nummer: 93-68-5
EINECS: 202-267-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR