(3-Fluoro-4-methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: 3-Fluoro-4-methoxyphenylacetic acid

M-Clarity™ quality level = MQ200

8.43872.0010 8.43872.0005
8.43872.0010EA 1120 NOK
8.43872.0010 8.43872.0005
(3-Fluoro-4-methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
(3-Fluoro-4-methoxyphenyl)acetic acid
Formel: C₉H₉FO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00060353
CAS-nummer: 452-14-2

Bestill nå

Spesifikasjon Testresultater

Molar Mass 184.16 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR