Divinylbenzene (mixture of isomers) ≥80% stabilised, Sigma-Aldrich®

Leverandør: Merck
Warning


Stabilisering: stabilisert med 4-tert-Butylpyrokatekol
M-Clarity™ quality level = MQ200

8.43986.1000
8.43986.1000EA 1100 NOK
8.43986.1000
Divinylbenzene (mixture of isomers) ≥80% stabilised, Sigma-Aldrich®
Divinylbenzene (mixture of isomers)
Formel: C₁₀H₁₀
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01778656
CAS-nummer: 1321-74-0
EINECS: 215-325-5
FN: 3082
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Solubility 0.005 g/l (25 °C)
Melting Point -67 °C
Molar Mass 130.18 g/mol
Boiling Point 195 °C (1013 hPa)
Vapor Pressure 0.9 hPa (20 °C)
Flash Point 77 °C
Explosion Limit 1.1 - 6.2 %(V)
Density 0.91 g/cm3 (20 °C)
Ignition Point 470 °C
Synonyms DVB
Hill Formula C10H10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR