Dimethyl succinate for syntese, Sigma-Aldrich®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

8.44002.0100 8.44002.1000
8.44002.0100EA 330 NOK
8.44002.0100 8.44002.1000
Dimethyl succinate for syntese, Sigma-Aldrich®
Dimethyl succinate
Formel: C₆H₁₀O₄
Kokepunkt: 200 °C (1013 hPa)
Smeltepunkt: 18…19 °C
Tetthet: 1,117 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008466
CAS-nummer: 106-65-0
EINECS: 203-419-9
Merck Indeks: 12,09038

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR