Lucifer Yellow Cadaverine fluorescent dye, for histology

Leverandør: Biotium

80018
80018.EA 2450 NOK
80018.
Lucifer Yellow Cadaverine fluorescent dye, for histology
Lucifer Yellow Cadaverine

Like Lucifer Yellow CH, Lucifer Yellow Cadaverine can be used as a fixable fluorescent tracer. In addition, the dye can be used to conjugate to carboxylic acid groups.


  • λEx/λEm= 428/536 nm
  • Yellow solid soluble in water
  • Store at 4°C and protect from light

Formel: C₁₇H₁₇K₂N₃O₈S₂
MW: 533.66 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 149733-79-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR