Silikagel Rubin drying bags, crystalline 1.0-3.0 mm, with indicator 1/3 TE ( ~10 g/bag), Fluka™

Leverandør: Honeywell Chemicals

80148-1KG
80148-1KGEA 3250 NOK
80148-1KG
Silikagel Rubin drying bags, crystalline 1.0-3.0 mm, with indicator 1/3 TE ( ~10 g/bag), Fluka™
Silikagel Rubin

With indicator function, 1/3 TE, bag with ~10 g, crystalline

Formel: SiO₂
MW: 60.08 g/mol
Kokepunkt: 2.230 °C
Smeltepunkt: 1.600 °C
Tetthet: 2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 112926-00-8
EINECS: 231-545-4

Bestill nå

Spesifikasjon Testresultater

Appearance of the substance complying
Sieve analysis 1-3 mm Min. 95 %
Water absorbing capacity Min. 20 %
Suitability as indicator complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR