Sodium polytungstate ~85% in water for separation of minerals mixtures, Supelco®

Leverandør: Merck
Danger

Synonymer: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate

80656-100ML
80656-100MLEA 5900 NOK
80656-100ML
Sodium polytungstate ~85% in water for separation of minerals mixtures, Supelco®
Sodium metatungstate monohydrate
Formel: Na₆O₃₉W₁₂·H₂O
MW: 2986.13 g/mol (2968.12 anhy)
MDL nummer: MFCD00061466
CAS-nummer: 12141-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=293cb9e4a121317ce26fb5d0a45022a54fcb8eb3&vid=7538771669f57ab581d62e120a37671bec60544b&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR