Pyridin, BAKER, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: Azabenzene, Azine

8074.2500EA 0 NOK
8074.2500
Pyridin, BAKER, Laboratory Reagent, J.T.Baker®
Pyridine
Formel: C₅H₅N
Kokepunkt: 115 °C (1013 hPa)
Smeltepunkt: –44 °C
Tetthet: 0,983 g/cm³ (20 °C)
Flash punkt: 17 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011732
CAS-nummer: 110-86-1
EINECS: 203-809-9
FN: 1282
ADR: 3,II
Merck Indeks: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Chloride (Cl) max. 0.005 %
Residue after Evaporation max. 0.005 %
Water (H2O) max. 0.25 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR