Natriumsulfat 99.0-101.0% (by titrimetric analysis) for HPLC, Supelco®

Leverandør: Merck

Synonymer: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

80948-250G
80948-250GEA 1220 NOK
80948-250G
Natriumsulfat 99.0-101.0% (by titrimetric analysis) for HPLC, Supelco®
Sodium sulphate
Formel: Na₂SO₄
MW: 142.04 g/mol
Kokepunkt: 1429
Smeltepunkt: 888
MDL nummer: MFCD00003504
CAS-nummer: 7757-82-6
Merck Indeks: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd15a8d25b473ddebcb8f8fb2df969474beccaf5&vid=ec998b618e6628bd41a34faeb5488913544b1bde&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR