Ammoniumhydrogendifluorid ≥98.5%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Danger

Synonymer: Ammonium bifluoride, Ammonium hydrogen fluoride

81000.260EA 1075 NOK
81000.260
Ammoniumhydrogendifluorid ≥98.5%, AnalaR NORMAPUR® analytical reagent
Ammonium hydrogen difluoride
Formel: NH₄HF₂
MW: 57.04 g/mol
Smeltepunkt: 127
Tetthet: 0.8 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012018
CAS-nummer: 1341-49-7
EINECS: 215-676-4
FN: 1727
ADR: 8 II
Merck Indeks: 13,00498

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98.5 %
Heavy metals (as Pb) Max. 0.03 %
Ignition residue (SO4) Max. 0.02 %
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 10 ppm
Fe (Iron) Max. 80 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR