Polystyrene standard ReadyCal set Mp 400-2000000, analytical standards for GPC
Leverandør: Merck
81434-1EA
81434-1EAEA 12925 NOK
81434-1EA
Polystyrene standard ReadyCal set Mp 400-2000000, analytical standards for GPC
Standarder kromatografi standarder
Set contains 12 different polystyrene standards with Mp ~374, ~1620, ~3420, ~9130, ~18100, ~32500, ~67500, ~128000, ~246000, ~659000, ~1000000, ~2180000; always 4 standards are lyophilised in a 1,5 ml vial (green, red or white); for 1 complete calibration use one of each of the green, red and white vials; 1 set contains 30 vials for 10 complete calibrations.
Sertifiseringer: Exact values can be taken from the accompanying certificate of analysis
NB: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.