Acetonitril, BAKER BIO-ANALYZED®, J.T.Baker®

Leverandør: Avantor
Danger

8144.4000GLEA 4400 NOK
8144.4000GL
Acetonitril, BAKER BIO-ANALYZED®, J.T.Baker®
Acetonitril
Formel: H₃CCN
Kokepunkt: 81,6 °C (1013 hPa)
Tetthet: 0,782 g/cm³ (20 °C)
Flash punkt: 2 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001878
CAS-nummer: 75-05-8
EINECS: 200-835-2
FN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Indeks: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (corrected for H2O) min. 99.9 %
Residue after Evaporation max. 1 ppm
Titrable Acid (µeq/g) max. 0.5
Titrable base (µeq/g) max. 0.6
Water (H2O) max. 30 ppm
Fluorescence Trace Impurities (as quinine base), ppb:
Measured at 450 nm max. 0.3
Measured at Emission Maximum for Solvent Impurities max. 1.0
Ultraviolet Absorbance (1.00-cm path vs water):
at 200 nm max. 0.05
at 220 nm max. 0.01
at 254-400 nm max. 0.01
UV Cut-off, nm max. 190


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR