Tape dispenser, stainless steel
Leverandør: ULTRATAPE
UT962
817-0311EA 18925 NOK
817-0311
Tape dispenser, stainless steel
Etikett- og tapedispensere
These dispensers are made from industrial 304 grade stainless steel with a mounting bracket on the bottom. It is corrosion resistant and the replaceable serrated blade is electroplated chrome buffed to a dull shine. It is designed to hold one roll, with a 76 mm core, up to 50,8 mm width and 201,2 metres in length.