Tape 4910, VHB™
Leverandør: 3M
4910
817-0447EA 970 NOK
817-0447
Tape 4910, VHB™
Tape
Clear, 1 mm thick, general-purpose tape with acrylic adhesive on both sides. It offers design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces. It is colourless, making it ideal for bonding transparent materials or for applications where a coloured bond line is unacceptable.
  • Fast and easy-to-use permanent bonding method provides high strength and long-term durability
  • Pressure sensitive adhesive bonds on contact to provide immediate handling strength
  • Virtually invisible fastening keeps surfaces smooth, even edges