Multi-roll tape dispenser, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H134680000
817-1390EA 6600 NOK
817-1390
Multi-roll tape dispenser, SP Bel-Art
Etikett- og tapedispensere
Write-On™ labeling tape or any tape with a 76 mm internal diameter will fit on the drum.
  • Mix colours with different widths
  • Flat writing surface helps make neater labels
  • Heavy steel base coated for easy cleaning and rubber feet to stop sliding
Bestillingsinformasjon: The tapes need to be ordered separately.