VWR®, Scientific Calculator
Leverandør: VWR Collection
818-0062EA 565 NOK
818-0062
VWR®, Scientific Calculator
Kalkulator Lommekalklulator
Convenient pocket size calculator can handle 99% of all routine and other calculations in the laboratory. Provides immediate, exact results for a wide range of applications: Conversions, scientific calculations, statistical operations, arithmetic calculations and similar laboratory work. Display: 10 characters with two-digit exponent.
  • Clear display with large, bright numbers and ergonomic keys
  • Three key memory and memory protection when the calculator is switched off
  • Automatic switch-off to preserve batteries
Functions: Over 50 functions forMore Product Information