Laboratorienotatbøker for renrom, TexWrite® 22
Leverandør: Texwipe
TX5708
818-0247EA 25050 NOK
818-0247
Laboratorienotatbøker for renrom, TexWrite® 22
Notatbøker
TexWrite® 22 Cleanroom Laboratory Notebooks are the only notebooks available made with synthetic cleanroom bond stationery. Cleanroom stationery is ideal in critical environments where very low particle and fibre generation is important. The pages of the notebook are sewn and bound with a Mylar®-sealed cover. The notebooks are cleaned and double packaged for cleanroom use. An index page is provided and each page is individually numbered for data retrieval ease. All pages contain dual signature lines and the pages are not removable. These features provide security to support Good LaboratoryMore Product Information