VWR®, Cleanroom Spiral Notebooks
Leverandør: VWR Collection
818-0279EA 2700 NOK
818-0279 818-0278 818-0280
VWR®, Cleanroom Spiral Notebooks
Notatbøker
These cleanroom spiral notebooks are made of synthetic latex paper.
  • White
  • 100 pages
  • Polypropylene cover
  • Spiral bound
  • 85 gsm
Suitable for cleanroom use.