1-Propanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis) for ion pair chromatography, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

81806-10G-F
81806-10G-FEA 8310 NOK
81806-10G-F
1-Propanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis) for ion pair chromatography, Supelco®
1-Propanesulphonic acid sodium salt monohydrate
Formel: CH₃CH₂CH₂SO₃Na·H₂O
MW: 164.16 g/mol
MDL nummer: MFCD00167033
CAS-nummer: 304672-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e00ca3d759843b03080b0e7ded357bc6bf182ae&vid=80b0b2723f3f214228c9fc75f97b7b18335a9f4a&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR