Anti-static cleanroom tapes, NovaStat™
Leverandør: MICRONOVA
ASTPC-272_U ASTPC-3436
819-0073EA 0 NOK
819-0073 819-0017
Anti-static cleanroom tapes, NovaStat™
Tape
These anti-static cleanroom tapes are made of cellophane with rubber based adhesive and are specially designed for use in ESD-sensitive environments.
  • Suitable for sealing and packaging
  • Will not create a static charge
  • Wound on a 3" plastic core
Colour: Amber transparent