Repositionable spray adhesive, 75
Leverandør: 3M
75
819-0122EA 7475 NOK
819-0122
Repositionable spray adhesive, 75
Klebemidler
3M™ repositionable spray adhesive 75 is a versatile, pressure sensitive, adhesive spray suitable for temporary holding or positioning of lightweight materials for display or prior to final assembly. It adheres in seconds, yet has an extra-long tack range that allows lifting and repositioning of materials.
  • Hold and position lightweight materials temporarily
  • Low soak-in formula allows it to bond delicate substrates without damaging the material
  • Formula resists bleeding and staining for reliable and effective performance
  • Dries clear, produces a smooth glue