Packaging tape
Leverandør: DESCO
242210
819-0142EA 110 NOK
819-0142
Packaging tape
Tape
Desco Europe packaging tape is a conventional self-adhesive PVC tape.
  • Suitable for sealing secondary packaging such as cartons
  • Conventional PVC self adhesive yellow tape, not for use in an EPA
  • Printed black with logo and text to EN 61340-5
It uses an acrylic gum adhesive. The tape is yellow and is printed in black with the ESD Sensitivity Symbol ( Ref: EN 61340-5-1) alerting that package contents are ESD sensitive and should be opened only by a grounded operator at an ESD protective workstation.
The tape is tribocharging and is not suitable for useMore Product Information