2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Danger

8235.2500EA 2525 NOK
8235.2500 8235.1000
2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®
2-Propanol
Formel: (CH₃)₂CHOH
Kokepunkt: 82 °C (1013 hPa)
Smeltepunkt: –89 °C
Tetthet: 0,786 g/cm³ (20 °C)
Flash punkt: 12 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011674
CAS-nummer: 67-63-0
EINECS: 200-661-7
FN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Indeks: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Exceeds ACS Specifications
Assay (by GC) (corrected for water) min. 99.5 %
Carbonyl Compounds (max. 0.002% Propionaldehyde and max. 0.002% Acetone) passes test
Color (APHA) max. 10
Residue after Evaporation max. 5 ppm
Solubility in Water passes test
Titrable Acid or Base (meq/g) max. 0.0001
Water (H2O) max. 0.05 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 210 nm max. 1.00
at 220 nm max. 0.40
at 230 nm max. 0.20
at 245 nm max. 0.08
at 260 nm max. 0.04
at 275 nm max. 0.03
at 300 nm max. 0.02
at 330-400 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR