Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC

Leverandør: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

83622.320EA 2925 NOK
83622.320
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Iso-Hexane
Formel: C₆H₁₄
MW: 86.18 g/mol
Smeltepunkt: –154 °C
Tetthet: 0.653 g/cm³ (25 °C)
Flash punkt: –33 °C
Lagringstemperatur: Romtemperatur
CAS-nummer: 64742-49-0
EINECS: 295-570-2
FN: 1208
ADR: 3 II
REACH: 01-2119484651-34
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce61b1646b637970765dc4b16a997650aeccfd1&vid=71606860b34952f73fd29b7df735dd0916b1e93c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless liquid
Assay (Mix. of C₆H₁₄ isomers) Min. 97.00 %
Water (KF) Max. 0.0100 % w/w
Residue after evaporation Max. 0.0005 % w/w
n-Hexane Max. 3.0 %
Transmittance (205 nm) Min. 50.0 %
Transmittance (220 nm) Min. 80.0 %
Transmittance (230 nm) Min. 95.0 %
Transmittance (280 nm) Min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR