Toluen ≥99.8%, HiPerSolv CHROMANORM® for HPLC

Leverandør: VWR Chemicals
Danger

Synonymer: Methylbenzene, Phenylmethane, FM approval

Filtered through a 0.2 µm filter, packaged under nitrogen.

83625.320EA 1375 NOK
83625.320 83625.290
Toluen ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Toluen
Formel: C₆H₅CH₃
MW: 92.14 g/mol
Kokepunkt: 110.6 °C (1013 hPa)
Smeltepunkt: –95 °C
Tetthet: 0.867 g/cm³ (20 °C)
Flash punkt: 4
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008512
CAS-nummer: 108-88-3
EINECS: 203-625-9
FN: 1294
ADR: 3 II
REACH: 01-2119471310-51
Merck Indeks: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) Min. 99,8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.05 %
Absorbance (300 nm) Max. 0.15
Absorbance (310 nm) Max. 0.1
Absorbance (330 nm) Max. 0.02
Absorbance (350 nm) Max. 0.01
Transmittance (300 nm) Min. 70 %
Transmittance (310 nm) Min. 80 %
Transmittance (330 nm) Min. 95 %
Transmittance (350 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR