Metanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments

Leverandør: VWR Chemicals
Danger

Synonymer: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.2 µm filter, packaged under nitrogen.

83638.320EA 513 NOK
83638.320 83638.290
Metanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Metanol
Formel: CH₃OH
MW: 32.04 g/mol
Kokepunkt: 64.6 °C (1013 hPa)
Smeltepunkt: –98 °C
Tetthet: 0.7918 g/cm³ (20 °C)
Flash punkt: 11 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004595
CAS-nummer: 67-56-1
EINECS: 200-659-6
FN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Indeks: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) Min. 99.9 %
Carbonyl compounds Passes test
Acidity Max. 0.0002 Meq/g
Alkalinity Max. 0.0002 Meq/g
Density (20/20) 0.791 --> 0.793
Evaporation residue Max. 3 ppm
Water Max. 0.03 %
Ca (Calcium) Max. 200 ppb
K (Potassium) Max. 100 ppb
Mg (Magnesium) Max. 100 ppb
Na (Sodium) Max. 200 ppb
Gradient (235 nm) Max. 2 MAU
Gradient (254 nm) Max. 2 MAU
Transmittance (205 nm) Min. 10 %
Transmittance (210 nm) Min. 40 %
Transmittance (220 nm) Min. 60 %
Transmittance (230 nm) Min. 80 %
Transmittance (240 nm) Min. 90 %
Transmittance (from 260 nm) Min. 98 %
Fluorescence (as quinine) (254 nm) Max. 1 ppb
Fluorescence (as quinine) (365 nm) Max. 1 ppb
Conforms to ACS Passes test
Filtered through 0.2 μm Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR