Sykloheksan ≥99.5%, PESTINORM® for pesticide residue analysis

Leverandør: VWR Chemicals
Danger

Synonymer: Hexahydrobenzene, Hexamethylene

Filtered through a 0.2 µm filter, packaged under nitrogen.

83658.320EA 1550 NOK
83658.320 83658.290 83658.400
Sykloheksan ≥99.5%, PESTINORM® for pesticide residue analysis
Sykloheksan
Formel: C₆H₁₂
MW: 84.16 g/mol
Kokepunkt: 81
Smeltepunkt: 7
Tetthet: 0.7785 g/cm³ (20 °C)
Flash punkt: –18 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003814
CAS-nummer: 110-82-7
EINECS: 203-806-2
FN: 1145
ADR: 3 II
REACH: 01-2119463273-41
Merck Indeks: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (on anhydrous substance) Min. 99.5 %
Evaporation residue Max. 5 ppm
Water Max. 0.02 %
Pesticide analysis (Lindane/ECD) Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR