Dimetylsulfoksid, dehydrert (maks. 0,005% H₂O) ≥99.8%

Leverandør: VWR Chemicals


0,2 µm Filtrert. Packaged under nitrogen.

83673.230EA 630 NOK
83673.230 83673.290
Dimetylsulfoksid, dehydrert (maks. 0,005% H₂O) ≥99.8%
Dimetylsulfoksid

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Formel: (CH₃)₂SO
Kokepunkt: 189 °C (1013 hPa)
Tetthet: 1,101 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
REACH: 01-2119431362-50
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (on anhydrous substance) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Residue on evaporation Max. 0.0005 %
Water Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR