Natriumsulfid hydrat, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

Synonymer: Dinatriumsulfid hydrat

83757.290EA 1175 NOK
83757.290
Natriumsulfid hydrat, GPR RECTAPUR®
Natriumsulfid hydrat
Formel: Na₂S·~3H₂O
MW: 96.06 g/mol (anhy)
Kokepunkt: ~920 °C (1013 hPa)
Smeltepunkt: 50
Tetthet: ∼1.43 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00149183
CAS-nummer: 27610-45-3
EINECS: 215-211-5
FN: 1849
ADR: 8 II
REACH: 01-2119513694-38
Merck Indeks: 12,08829

Bestill nå

Spesifikasjon Testresultater

Assay 58 - 64 %
SO3 (Sulphite) Max. 0.6 %
S2O3 (Thiosulphate) Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR