Saltsyre 34%, NORMATOM® for trace metal analysis
Leverandør: VWR Chemicals
Danger
Synonymer: Hydrochloric acid solution, Muriatic acid
83871.270EA 1590 NOK
83871.270 83871.330 83871.290
Saltsyre 34%, NORMATOM® for trace metal analysis
Saltsyre
Formel: HCl
Kokepunkt: 110 °C (1013 hPa)
Smeltepunkt: –30 °C
Tetthet: 1,18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,II
REACH: 01-2119484862-27
Bestill nå
Spesifikasjon Testresultater
Assay 34 - 37 %
Colouration Max. 10 APHA
Br (Bromide) Max. 10 ppm
Total P (Phosphorus) Max. 0.01 ppm
Total S (Sulphur) Max. 0.30 ppm
Ag (Silver) Max. 1 ppb
Al (Aluminium) Max. 1 ppb
As (Arsenic) Max. 0.5 ppb
Au (Gold) Max. 0.5 ppb
B (Boron) Max. 1 ppb
Ba (Barium) Max. 0.1 ppb
Be (Beryllium) Max. 0.1 ppb
Bi (Bismuth) Max. 0.1 ppb
Ca (Calcium) Max. 1 ppb
Cd (Cadmium) Max. 0.1 ppb
Ce (Cerium) Max. 0.1 ppb
Co (Cobalt) Max. 0.1 ppb
Cr (Chromium) Max. 0.5 ppb
Cs (Cesium) Max. 0.1 ppb
Cu (Copper) Max. 0.5 ppb
Dy (Dysprosium) Max. 0.1 ppb
Er (Erbium) Max. 0.1 ppb
Eu (Europium) Max. 0.1 ppb
Fe (Iron) Max. 1 ppb
Ga (Gallium) Max. 0.1 ppb
Gd (Gadolinium) Max. 0.1 ppb
Ge (Germanium) Max. 0.1 ppb
Hf (Hafnium) Max. 0.1 ppb
Hg (Mercury) Max. 0.1 ppb
Ho (Holmium) Max. 0.1 ppb
In (Indium) Max. 0.1 ppb
K (Potassium) Max. 1 ppb
La (Lanthanum) Max. 0.1 ppb
Li (Lithium) Max. 0.1 ppb
Lu (Lutetium) Max. 0.1 ppb
Mg (Magnesium) Max. 0.5 ppb
Mn (Manganese) Max. 0.1 ppb
Mo (Molybdenum) Max. 0.1 ppb
Na (Sodium) Max. 1 ppb
Nb (Niobium) Max. 0.1 ppb
Nd (Neodymium) Max. 0.1 ppb
Ni (Nickel) Max. 0.5 ppb
Pb (Lead) Max. 0.1 ppb
Pr (Praseodymium) Max. 0.1 ppb
Rb (Rubidium) Max. 0.1 ppb
Re (Rhenium) Max. 0.1 ppb
Rh (Rhodium) Max. 0.5 ppb
Ru (Ruthenium) Max. 0.1 ppb
Sb (Antimony) Max. 0.5 ppb
Sc (Scandium) Max. 0.100 ppb
Se (Selenium) Max. 1 ppb
Sm (Samarium) Max. 0.100 ppb
Sn (Tin) Max. 0.500 ppb
Sr (Strontium) Max. 0.100 ppb
Tb (Terbio) Max. 0,100 ppb
Te (Tellurium) Max. 0.100 ppb
Th (Thorium) Max. 0.100 ppb
Ti (Titanium) Max. 0.500 ppb
Tl (Thallium) Max. 0.100 ppb
Tm (Thulium) Max. 0.100 ppb
U (Uranium) Max. 0.100 ppb
V (Vanadium) Max. 0.500 ppb
W (Tungsten) Max. 0.100 ppb
Y (Yttrium) Max. 0.100 ppb
Yb (Ytterbium) Max. 0.100 ppb
Zn (Zinc) Max. 1 ppb
Zr (Zirconium) Max. 0.100 ppb