Salpetersyre 67%, NORMATOM® for trace metal analysis

Leverandør: VWR Chemicals
Danger

Synonymer: Nitric acid solution

83872.270EA 960 NOK
83872.270 83872.330 83872.290
Salpetersyre 67%, NORMATOM® for trace metal analysis
Salpetersyre
Formel: HNO₃
Kokepunkt: 120,5 °C (1013 hPa)
Smeltepunkt: –42 °C
Tetthet: 1,4 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
FN: 2031
ADR: 8 (5.1),II
Merck Indeks: 13,06608

Bestill nå

Spesifikasjon Testresultater

Assay 67 - 69 %
Colouration Max. 10 APHA
Cl (Chloride) Max. 0.2 ppm
Total P (Phosphorus) Max. 0.01 ppm
Total S (Sulphur) Max. 0.3 ppm
Ag (Silver) Max. 0.1 ppb
Al (Aluminium) Max. 1 ppb
As (Arsenic) Max. 0.5 ppb
Au (Gold) Max. 0.1 ppb
B (Boron) Max. 1 ppb
Ba (Barium) Max. 0.1 ppb
Be (Beryllium) Max. 0.1 ppb
Bi (Bismuth) Max. 0.1 ppb
Ca (Calcium) Max. 1 ppb
Cd (Cadmium) Max. 0.5 ppb
Ce (Cerium) Max. 0.1 ppb
Co (Cobalt) Max. 0.5 ppb
Cr (Chromium) Max. 1 ppb
Cs (Cesium) Max. 0.1 ppb
Cu (Copper) Max. 0.5 ppb
Dy (Dysprosium) Max. 0.1 ppb
Er (Erbium) Max. 0.1 ppb
Eu (Europium) Max. 0.1 ppb
Fe (Iron) Max. 1 ppb
Ga (Gallium) Max. 0.1 ppb
Gd (Gadolinium) Max. 0.1 ppb
Ge (Germanium) Max. 0.1 ppb
Hf (Hafnium) Max. 0.1 ppb
Hg (Mercury) Max. 0.1 ppb
Ho (Holmium) Max. 0.1 ppb
In (Indium) Max. 0.1 ppb
K (Potassium) Max. 1 ppb
La (Lanthanum) Max. 0.1 ppb
Li (Lithium) Max. 0.1 ppb
Lu (Lutetium) Max. 0.1 ppb
Mg (Magnesium) Max. 1 ppb
Mn (Manganese) Max. 0.1 ppb
Mo (Molybdenum) Max. 0.1 ppb
Na (Sodium) Max. 1 ppb
Nb (Niobium) Max. 0.1 ppb
Nd (Neodymium) Max. 0.1 ppb
Ni (Nickel) Max. 0.5 ppb
Pb (Lead) Max. 0.1 ppb
Pd (Palladium) Max. 0.5 ppb
Pr (Praseodymium) Max. 0.1 ppb
Pt (Platinum) Max. 0.5 ppb
Rb (Rubidium) Max. 0.1 ppb
Re (Rhenium) Max. 0.1 ppb
Rh (Rhodium) Max. 0.5 ppb
Ru (Ruthenium) Max. 0.500 ppb
Sb (Antimony) Max. 0.500 ppb
Sc (Scandium) Max. 0.100 ppb
Se (Selenium) Max. 1 ppb
Sm (Samarium) Max. 0.100 ppb
Sn (Stagno) Max. 0,500 ppb
Sr (Strontium) Max. 0.100 ppb
Tb (Terbium) Max. 0.100 ppb
Te (Tellurium) Max. 0.100 ppb
Th (Thorium) Max. 0.100 ppb
Ti (Titanium) Max. 0.500 ppb
Tl (Thallium) Max. 0.100 ppb
Tm (Thulium) Max. 0.100 ppb
U (Uranium) Max. 0.100 ppb
V (Vanadium) Max. 0.500 ppb
W (Tungsten) Max. 0.100 ppb
Y (Yttrium) Max. 0.100 ppb
Yb (Ytterbium) Max. 0.100 ppb
Zn (Zinc) Max. 0.500 ppb
Zr (Zirconium) Max. 0.100 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR