Supelco® Colour Reference Solutions, set of colours Y, ampoules of 2 ml, Merck

Leverandør: Merck

83883-1SET-F
83883-1SET-FEA 3775 NOK
83883-1SET-F
Supelco® Colour Reference Solutions, set of colours Y, ampoules of 2 ml, Merck
Standarder Fargereferanse og spekroskopistandarer

The kit contains a set of seven yellow colour reference standards (identified sequentially from Y1 to Y7) and a 1% hydrogen chloride solution in water. The solutions are produced from two primary metal salt solutions of ferric(III) chloride and cobalt(II) chloride, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).


  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Available in tamper-evident 2 ml ampoules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system


They are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red colour range.


This is a set of the following color reference solutions according to PhEur:

Colour Reference Solution Y1 - 2 ml ampoule
Colour Reference Solution Y2 - 2 ml ampoule
Colour Reference Solution Y3 - 2 ml ampoule
Colour Reference Solution Y4 - 2 ml ampoule
Colour Reference Solution Y5 - 2 ml ampoule
Colour Reference Solution Y6 - 2 ml ampoule
Colour Reference Solution Y7 - 2 ml ampoule
Blind sample : Hydrochloric acid solution, 1% in water - 2 ml ampoule


NB: Contains Cobalt(II) chloride.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR