n-Pentan ≥95%, HiPerSolv CHROMANORM® for HPLC

Leverandør: VWR Chemicals
Danger

Synonymer: Pentane

Filtered through a 0.2 µm filter, packaged under nitrogen.

83993.320EA 2250 NOK
83993.320
n-Pentan ≥95%, HiPerSolv CHROMANORM® for HPLC
n-Pentane
Formel: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kokepunkt: 35…36 °C (1013 hPa)
Smeltepunkt: –130 °C
Tetthet: 0.63 g/cm³ (20 °C)
Flash punkt: –40 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009498
CAS-nummer: 109-66-0
EINECS: 203-692-4
FN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Indeks: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (on anhydrous substance) Min. 95.0 %
Acidity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.01 %
Transmittance (200 nm) Min. 56 %
Transmittance (210 nm) Min. 71 %
Transmittance (220 nm) Min. 89 %
Transmittance (230 nm) Min. 98 %
Transmittance (240 nm) Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR