Salpetersyre ≥65% (by titrimetric analysis), Puriss. p.a. (maks. 0,0000005% Hg), Fluka™

Leverandør: Honeywell Chemicals
Danger

84378-2.5L 84378-1L
84378-2.5LEA 1925 NOK
84378-2.5L 84378-1L
Salpetersyre ≥65% (by titrimetric analysis), Puriss. p.a. (maks. 0,0000005% Hg), Fluka™
Salpetersyre
Formel: HNO₃
Kokepunkt: 122 °C (1,013 hPa)
Smeltepunkt: –31 °C
Tetthet: 1,400…1.480 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
FN: 2031
ADR: 8,II
Merck Indeks: 13,06608

Bestill nå

Spesifikasjon Testresultater

Ignition Residue ≤0.0005% (as SO₄)
Assay 65.0 - 66.0 %
Density (D 20/4) 1.390 - 1.410
Residue on ignition (SO₄) Max. 5 ppm
Silver (Ag) Max. 0.01 ppm
Arsenic (As) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Barium (Ba) Max. 0.01 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Mercury (Hg) Max. 0.005 ppm
Potassium (K) Max. 0.05 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.01 ppm
Strontium (Sr) Max. 0.01 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.05 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR