Jod concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Leverandør: VWR Chemicals


Dose for preparation of 1 litre of solution.

84591.600EA 550 NOK
84591.600
Jod concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Jod
Formel: I₂
MW: 253.81 g/mol
Kokepunkt: 184.3
Smeltepunkt: 114
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011355
CAS-nummer: 7553-56-2
EINECS: 231-442-4
Merck Indeks: 13,05036

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.04998 - 0.05002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR