Natriumhydroksid concentrate for dilution to 0.01 mol aqueous solution, ConvoL NORMADOSE® Volumetric

Leverandør: VWR Chemicals
Danger

Synonymer: Caustic soda

conc_1l

84596.600EA 305 NOK
84596.600
Natriumhydroksid concentrate for dilution to 0.01 mol aqueous solution, ConvoL NORMADOSE® Volumetric
Natriumhydroksid
Formel: NaOH
MW: 40 g/mol
Kokepunkt: >105 °C (1013 hPa)
Smeltepunkt: 323
Tetthet: 1.327 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8 II
Merck Indeks: 14,08627

Bestill nå

Spesifikasjon Testresultater

Concentration (20°C; accuracy +/-2%) 0.0098 - 0.0102 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR