VWR Chemicals, Base Materials for Dehydrated Media
Leverandør: VWR Chemicals
26208.297EA 5525 NOK
26208.297 499332OO 84687.0500 84688.0500 84618.0500 84620.0500 84623.0500 84609.0500 84601.0500 84616.0500 84610.0500 84615.0500 84862.25000 84601.5000 84609.5000 84610.5000 84620.5000 84688.5000 85862.5000 85862.0500 84615.5000 85895.25000 85896.25000 85898.25000 85899.25000 92424.25000 92425.25000 92426.25000
VWR Chemicals, Base Materials for Dehydrated Media
Mikrobiologiske medier Base Materialer for Mikrobiologi
The ingredients used to prepare culture media are very important, because ultimately the results depend on the quality of these compounds. Raw materials should be stored at 10 to 30 °C unless otherwise stated, in their closed containers in a dry environment.
  • Ingredients are specifically selected to ensure consistent, reproducible results
  • Manufactured under the strictest cGMP standards