Toluen ≥99.9% (by GC), SPECTRONORM® for spectroscopy

Leverandør: VWR Chemicals
Danger

Synonymer: Methylbenzene, Phenylmethane, FM approval

Filtered through a 0.2 µm filter, packaged under nitrogen.

84708.290EA 885 NOK
84708.290
Toluen ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Toluen

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

Formel: C₆H₅CH₃
MW: 92.14 g/mol
Kokepunkt: 110.6 °C (1013 hPa)
Smeltepunkt: –95 °C
Tetthet: 0.867 g/cm³ (20 °C)
Flash punkt: 4 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008512
CAS-nummer: 108-88-3
EINECS: 203-625-9
FN: 1294
ADR: 3 II
REACH: 01-2119471310-51
Merck Indeks: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 99.9%
Acidity max. 0.0001 meq/g
Alkalinity max. 0.0002 meq/g
Evaporation residue max. 0.0002%
Water max. 0.01%
Color max. 10 Hz
Fluorescence (as quinine at 365 nm) max. 1.0 ppb
Transmission at 285 nm min. 15%
Transmission at 290 nm min. 60%
Transmission at 300 nm min. 80%
Transmission at 310 nm min. 90%
Transmission at 335 nm min. 96%
Transmission from 350 nm min. 98%
Absorbance at 285 nm max. 0.82
Absorbance at 290 nm max. 0.22
Absorbance at 300 nm max. 0.10
Absorbance at 310 nm max. 0.05
Absorbance at 335 nm max. 0.02
Absorbance from 350 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR