n-Heptan, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Leverandør: Avantor

Synonymer: high purity solvents

8472.2500EA 3050 NOK
8472.2500
n-Heptan, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
n-Heptane
Formel: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Kokepunkt: 98.4
Smeltepunkt: -90.6
Flash punkt: -4
MDL nummer: MFCD00009544
CAS-nummer: 142-82-5
FN: 1206
ADR: 3 II
Merck Indeks: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (by GC) min. 95.0 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR