Svovelsyre solution 49-51% (by titrimetric analysis) for HPLC, Fluka™

Leverandør: Honeywell Chemicals
Danger

84733-100ML 84733-500ML
84733-100MLEA 1250 NOK
84733-100ML 84733-500ML
Svovelsyre solution 49-51% (by titrimetric analysis) for HPLC, Fluka™
Svovelsyre
Formel: H₂SO₄
MW: 98.08 g/mol
Kokepunkt: 110 °C (1.013 hPa)
Smeltepunkt: 10.38
Tetthet: 1.290 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 2796
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Assay 49.0 - 51.0 %
Density (D 20/4) 1.375 - 1.405
Absorbance 500 nm Max. 0.02
Absorbance at 240 nm Max. 0.02
Absorbance at 230 nm Max. 0.03
Absorbance at 220 nm Max. 0.04
Absorbance at 210 nm Max. 0.05
Suitability test (HPLC) complying

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR