VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 33 Elements, for ICP
Leverandør: VWR Chemicals
84791.180EA 7325 NOK
84791.180
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 33 Elements, for ICP
Standarder Kationstandarder Multi-Element Standards
33 elements 100 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.