VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 9 Elements in Hydrochloric Acid 10% for ICP
Leverandør: VWR Chemicals
84792.180EA 12375 NOK
84792.180
VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 9 Elements in Hydrochloric Acid 10% for ICP
Standarder Kationstandarder Multi-Element Standards
9 Elements 100 mg/L: Au, Ir, Pd, Pt, Rh, Ru, Os, Sn, Te.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.