VWR® ARISTAR®, Multi-Element Quality Control Standard, 10 mg/L for 36 Elements in 2% Nitric Acid , for ICP-MS
Leverandør: VWR Chemicals
84793.180EA 5425 NOK
84793.180
VWR® ARISTAR®, Multi-Element Quality Control Standard, 10 mg/L for 36 Elements in 2% Nitric Acid , for ICP-MS
Standarder Kationstandarder Multi-Element Standards
36 Elements: 10 mg/L: Al, Ag, As, B, Ba, Ca, Cd, Ce, Co, Cr, Cu, Dy, Er, Eu, Fe, Gd, Ho, K, La, Li, Lu, Mg, Mn, Na, Nd, Ni, P, Pb, Rb, Se, Sm, Sr, Tl, Tm, V, Zn
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.