EDTA dinatrium salt concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Leverandør: VWR Chemicals

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

Dose for preparation of 1 litre of solution.

84864.600EA 480 NOK
84864.600
EDTA dinatrium salt concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
EDTA dinatrium salt

Dose for preparation of 1 litre of solution of concentration 0.1 mol/l

Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

Titer (20°C) 0.0995 - 0.1005 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR