Trietylamin ≥99.8%, HiPerSolv CHROMANORM® for LC-MS

Leverandør: VWR Chemicals
Danger

Synonymer: (Diethylamino)ethane, TEA

Packaged under nitrogen.

84883.180EA 2895 NOK
84883.180 84883.260
Trietylamin ≥99.8%, HiPerSolv CHROMANORM® for LC-MS
Trietylamin
Formel: (C₂H₅)₃N
MW: 101.19 g/mol
Kokepunkt: 90
Smeltepunkt: –115 °C
Tetthet: 0.729 g/cm³ (20 °C)
Flash punkt: –11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009051
CAS-nummer: 121-44-8
EINECS: 204-469-4
FN: 1296
ADR: 3 (8) II
Merck Indeks: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (on anhydrous substance) Min. 99.8 %
Evaporation residue Max. 0.0001 %
Water Max. 0.02 %
Transmittance (230 nm) (0.1 %) Min. 10 %
Transmittance (240 nm) (0.1 %) Min. 50 %
Transmittance (245 nm) (0.1 %) Min. 80 %
Transmittance (250 nm) (0.1 %) Min. 95 %
Transmittance (255 nm) (0.1 %) Min. 99 %
Suitable for LC-MS (0.1 %) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR