Methanol with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Leverandør: VWR Chemicals
Danger
84896.290EA 1250 NOK
84896.290
Methanol with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Methanol with 0,1% (v/v) formic acid
Flash punkt: 12 °C (closed cup)
Lagringstemperatur: Romtemperatur
FN: 1230
ADR: 3 (6.1) II
Bestill nå
Spesifikasjon Testresultater
Nominal concentration (V/V) 0.095 to 0.105 %
Evaporation residue Max. 0.0001 %
Water Max. 0.1 %
Ca (Calcium) Max. 0.2 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.2 ppm
Na (Sodium) Max. 0.5 ppm
Suitable for LC-MS Passes test
Transmittance (240 nm) Min. 50 %
Transmittance (250 nm) Min. 80 %
Transmittance (260 nm) Min. 95 %
Transmittance (270 nm) Min. 99 %