Benzoat 1.000 mg/l i vann enkeltgrunnstoffsstandard for ionekromatografi
Leverandør: VWR Chemicals
84955.180EA 1075 NOK
84955.180
Benzoat 1.000 mg/l i vann enkeltgrunnstoffsstandard for ionekromatografi
Standarder Anionstandarder Benzoate Standards
Sertifiseringer: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.