Iodat 1000 mg/l i vann enkeltgrunnstoffsstandard for ionekromatografi
Leverandør: VWR Chemicals
84971.180EA 900 NOK
84971.180
Iodat 1000 mg/l i vann enkeltgrunnstoffsstandard for ionekromatografi
Standarder Anionstandarder Iodate Standards
Sertifiseringer: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.