di-Litiumtetraborat ≥99.0%, AnalaR NORMAPUR® ACS for X-ray analysis

Leverandør: VWR Chemicals
Danger

85023.290EA 4475 NOK
85023.290 85023.360 85023.230
di-Litiumtetraborat ≥99.0%, AnalaR NORMAPUR® ACS for X-ray analysis
di-Litiumtetraborat
Formel: Li₂B₄O₇
Smeltepunkt: 918 °C
Tetthet: 1,4 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011083
CAS-nummer: 12007-60-2
EINECS: 234-514-3
Merck Indeks: 14,05524

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Bulk density (g/ml) Min. 0.25
Heavy metals (as Pb) Max. 10 ppm
Insoluble matter Max. 100 ppm
Loss on fusion (950°C) Max. 2.0 %
Phosphorus compounds (as PO4) Max. 40 ppm
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 100 ppm
Al (Aluminium) Max. 10 ppm
Ca (Calcium) Max. 30 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 30 ppm
Mg (Magnesium) Max. 5 ppm
Na (Sodium) Max. 50 ppm
Si (Silicium) Max. 80 ppm
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR